Dimethyl cyclopentylmalonate ≥97.0%

Supplier: TCI America

Synonyms: Cyclopentylmalonic Acid Dimethyl Ester, Dimethyl 2-cyclopentylmalonate

C1621-25G C1621-5G
TCC1621-025GEA 114 USD
TCC1621-025G TCC1621-5G
Dimethyl cyclopentylmalonate ≥97.0%
Dimethyl cyclopentylmalonate

CAS Number: 82491-60-9
MDL Number: MFCD03844781
Molecular Formula: C10H16O4
Molecular Weight: 200.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: Cyclopentylmalonic Acid Dimethyl Ester

Formula: C₁₀H₁₆O₄
Density: 1.09 (20 °C)
MDL Number: MFCD03844781
CAS Number: 82491-60-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94fab93ef5c2dd29d490afe6cb2beff9545e551f&vid=94fab93ef5c2dd29d490afe6cb2beff9545e551f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR