Benzyl-4-nitrophenyl carbonate ≥96.0%
Supplier: TCI America
CAS Number: 13795-24-9
MDL Number: MFCD00007323
Molecular Formula: C14H11NO5
Molecular Weight: 273.24
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 76
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Polyamine Synthesis Reagents
Other Product Names / Synonyms: Carbonic Acid Benzyl 4-Nitrophenyl Ester
Formula:
C₁₄H₁₁NO₅ Melting Pt: 76 °C |
MDL Number:
MFCD00007323 CAS Number: 13795-24-9 |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...