Captopril ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C1580-25G C1580-5G
TCC1580-025GEA 388 USD
TCC1580-025G TCC1580-005G
Captopril ≥98.0% (by HPLC, titration analysis)
Captopril

CAS Number: 62571-86-2
MDL Number: MFCD00168073
Molecular Formula: C9H15NO3S
Molecular Weight: 217.28
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 106
Specific rotation [a]20/D: -127.5 deg (C=1, EtOH)


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antihypertensive Ingredients, Prodrug Ingredients
Other Product Names / Synonyms: (S)-(-)-1-(3-Mercapto-2-methyl-1-oxopropyl)-L-proline, (S)-(-)-1-(3-Mercapto-2-methylpropionyl)-L-proline

Formula: C₉H₁₅NO₃S
Melting Pt: 106 °C
MDL Number: MFCD00168073
CAS Number: 62571-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ac7ad82d87f4b4d328cb36d2be6735e870df0f2&vid=4ac7ad82d87f4b4d328cb36d2be6735e870df0f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR