2,2-Dimethylcyclopropyl cyanide ≥97.0%
Supplier: TCI America
Synonyms:
1-Cyano-2,2-dimethylcyclopropane, 2,2-Dimethylcyclopropanecarbonitrile
C1524-1G
C1524-5G
TCC1524-001GEA
179
USD
TCC1524-001G
TCC1524-5G
2,2-Dimethylcyclopropyl cyanide ≥97.0%
2,2-Dimethylcyclopropyl cyanide
CAS Number: 5722-11-2
MDL Number: MFCD01321153
Molecular Formula: C6H9N
Molecular Weight: 95.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: 1-Cyano-2,2-dimethylcyclopropane
Formula:
C₆H₉N MW: 95.15 g/mol Density: 0.86 (20 °C) |
MDL Number:
MFCD01321153 CAS Number: 5722-11-2 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...