(S)-(+)-Camptothecin ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
(S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
C1495-1G
C1495-100MG
TCC1495-001GEA
342
USD
TCC1495-001G
TCC1495-100MG
(S)-(+)-Camptothecin ≥97.0% (by HPLC)
(S)-(+)-Camptothecin
CAS Number: 7689-03-4
MDL Number: MFCD00081076
Molecular Formula: C20H16N2O4
Molecular Weight: 348.36
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 40 deg (C=0.4, CHCl3:MeOH=8:2)
Formula:
C₂₀H₁₆N₂O₄ MW: 348.36 g/mol |
MDL Number:
MFCD00081076 CAS Number: 7689-03-4 |
Specification Test Results
Purity/Analysis Method | >97.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...