Chlorodiisopropylsilane ≥95.0%
Supplier: TCI America
Synonyms:
Diisopropylchlorosilane
C1492-5ML
C1492-25ML
TCC1492-005MLEA
107
USD
TCC1492-005ML
TCC1492-25ML
Chlorodiisopropylsilane ≥95.0%
Chlorodiisopropylsilane
CAS Number: 2227-29-4
MDL Number: MFCD00054896
Molecular Formula: C6H15ClSi
Molecular Weight: 150.72
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 143
Flash Point (°C): 22
Specific Gravity (20/20): 0.89
Product Categories: Synthetic Organic Chemistry, Reduction, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Diisopropylchlorosilane
Formula:
C₆H₁₅ClSi MW: 150.72 g/mol Boiling Pt: 143 °C Density: 0.89 (20 °C) Flash Pt: 22 °C |
MDL Number:
MFCD00054896 CAS Number: 2227-29-4 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...