1-(2-Chloroethyl)naphthalene ≥95.0%

Supplier: TCI America

C1490-5G C1490-25G
TCC1490-005GEA 175.8 USD
TCC1490-005G TCC1490-025G
1-(2-Chloroethyl)naphthalene ≥95.0%
1-(2-Chloroethyl)naphthalene

CAS Number: 41332-02-9
MDL Number: MFCD01321149
Molecular Formula: C12H11Cl
Molecular Weight: 190.67
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Specific Gravity (20/20): 1.15

Formula: C₁₂H₁₁Cl
MW: 190.67 g/mol
Boiling Pt: 168 °C
Density: 1.15 (20 °C)
MDL Number: MFCD01321149
CAS Number: 41332-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5aebb0617a740e50211924143add879755637cc7&vid=5aebb0617a740e50211924143add879755637cc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR