Chlorodimethyl(3-phenylpropyl)silane ≥97.0%
Supplier: TCI America
C1488-5ML
C1488-25ML
TCC1488-005MLEA
143
USD
TCC1488-005ML
TCC1488-025ML
Chlorodimethyl(3-phenylpropyl)silane ≥97.0%
Chlorodimethyl(3-phenylpropyl)silane
CAS Number: 17146-09-7
MDL Number: MFCD00049281
Molecular Formula: C11H17ClSi
Molecular Weight: 212.79
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 103
Specific Gravity (20/20): 0.99
Formula:
C₁₁H₁₇ClSi MW: 212.79 g/mol Boiling Pt: 121 °C Density: 0.99 (20 °C) Flash Pt: 103 °C |
MDL Number:
MFCD00049281 CAS Number: 17146-09-7 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...