1-Chloro-2-methylpentane ≥98.0%
Supplier: TCI America
C1476-5G
C1476-25G
TCC1476-005GEA
28
USD
TCC1476-005G
TCC1476-25G
1-Chloro-2-methylpentane ≥98.0%
1-Chloro-2-methylpentane
CAS Number: 14753-05-0
MDL Number: MFCD00053236
Molecular Formula: C6H13Cl
Molecular Weight: 120.62
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Specific Gravity (20/20): 0.88
Formula:
C₆H₁₃Cl MW: 120.62 g/mol Boiling Pt: 120 °C Density: 0.88 (20 °C) |
MDL Number:
MFCD00053236 CAS Number: 14753-05-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...