(S)-(-)-Citronellal ≥96.0%
Supplier: TCI America
Synonyms:
(S)-(-)-3,7-Dimethyl-6-octenal, (S)-(-)-3,7-Dimethyloct-6-enal, (-)-Citronellal, (S)-3,7-Dimethyloct-6-enal
C1454-25ML
C1454-5ML
TCC1454-025MLEA
188.19
USD
TCC1454-025ML
76219-674
(S)-(-)-Citronellal ≥96.0%
(S)-(-)-Citronellal
CAS Number: 5949-05-3
MDL Number: MFCD00075588
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 75
Specific Gravity (20/20): 0.85
Specific rotation [a]20/D: -15.5 deg (neat)
Product Categories: Bioscience, Acyclic Monoterpenes
Other Product Names / Synonyms: (S)-(-)-3,7-Dimethyl-6-octenal
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 84 °C (1013 hPa) Melting Pt: –63 °C Density: 0.85 (20 °C) Flash Pt: 75 °C |
MDL Number:
MFCD00075588 CAS Number: 5949-05-3 EINECS: 227-707-9 |
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...