Amyl acetoacetate ≥98.0%
Supplier: TCI America
Synonyms:
Acetoacetic Acid Pentyl Ester, Acetoacetic Acid Amyl Ester, Pentyl Acetoacetate
A0811-25ML
A0811-500ML
TCA0811-025MLEA
34.58
USD
TCA0811-025ML
TCA0811-500ML
Amyl acetoacetate ≥98.0%
Amyl acetoacetate
CAS Number: 6624-84-6
MDL Number: MFCD00059433
Molecular Formula: C9H16O3
Molecular Weight: 172.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 222
Specific Gravity (20/20): 0.96
Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Acetoacetic Acid Pentyl Ester, Acetoacetic Acid Amyl Ester, Pentyl Acetoacetate
Formula:
C₉H₁₆O₃ MW: 172.22 g/mol Boiling Pt: 222 °C Density: 0.96 (20 °C) |
MDL Number:
MFCD00059433 CAS Number: 6624-84-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...