(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (1R,2R)-Cyclohexane-1,2-diamine (2R,3R)-2,3-dihydroxysuccinate, (1R)-trans-1,2-Diaminocyclohexane L-Tartrate

C1449-5G C1449-25G
TCC1449-005GEA 26 USD
TCC1449-005G TCC1449-025G
(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate ≥98.0% (by titrimetric analysis)
(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate

CAS Number: 39961-95-0
MDL Number: MFCD00191979
Molecular Formula: C6H14N2
Molecular Weight: 264.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 273
Specific rotation [a]20/D: 12 deg (C=2.5, H2O)


Other Product Names / Synonyms: (1R)-trans-1,2-Diaminocyclohexane L-Tartrate

Formula: C₆H₁₄N₂
MW: 264.28 g/mol
Melting Pt: 273 °C
MDL Number: MFCD00191979
CAS Number: 39961-95-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45c503f5a98af5054e8748a4dc0f50825ac892a5&vid=45c503f5a98af5054e8748a4dc0f50825ac892a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR