4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine-1-oxyl ≥96.0% (by HPLC) free radical

Supplier: TCI America

Synonyms: 4-(2-Chloroacetamido)-TEMPO

C1432-1G C1432-100MG
TCC1432-1GEA 282 USD
TCC1432-1G TCC1432-100MG
4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine-1-oxyl ≥96.0% (by HPLC) free radical
4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine-1-oxyl

CAS Number: 36775-23-2
MDL Number: MFCD00041858
Molecular Formula: C11H20ClN2O2
Molecular Weight: 247.74
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Melting point (°C): 117


Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-(2-Chloroacetamido)-TEMPO

Formula: C₁₁H₂₀ClN₂O₂
MW: 247.74 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00041858
CAS Number: 36775-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f18c07584e7fa1cd471d913a2077dfacd3598153&vid=f18c07584e7fa1cd471d913a2077dfacd3598153&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR