2-(2-(2-Chloroethoxy)ethoxy)ethanol ≥96.0%

Supplier: TCI America
Warning

C1416-100G C1416-25G
TCC1416-100GEA 165.41 USD
TCC1416-100G TCC1416-025G
2-(2-(2-Chloroethoxy)ethoxy)ethanol ≥96.0%
2-(2-(2-Chloroethoxy)ethoxy)ethanol

CAS Number: 5197-62-6
MDL Number: MFCD00002874
Molecular Formula: C6H13ClO3
Molecular Weight: 168.62
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 232
Flash Point (°C): 107
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: Triethylene Glycol Monochlorohydrin

Formula: C₆H₁₃ClO₃
MW: 168.62 g/mol
Boiling Pt: 232 °C
Density: 1.16 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00002874
CAS Number: 5197-62-6
EINECS: 225-984-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fd8a1f533dc3cd36a331fbfc99ff6ac75a4aa02&vid=7fd8a1f533dc3cd36a331fbfc99ff6ac75a4aa02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR