4-Chloro-7-chlorosulfonyl-2,1,3-benzoxadiazole ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Chloro-7-chlorosulphonylbenzofurazan, 4-Chloro-2,1,3-benzoxadiazole-7-sulphonyl Chloride

C1394-1G C1394-5G
TCC1394-001GEA 104 USD
TCC1394-001G TCC1394-5G
4-Chloro-7-chlorosulfonyl-2,1,3-benzoxadiazole ≥97.0% (by titrimetric analysis)
4-Chloro-7-chlorosulfonyl-2,1,3-benzoxadiazole

[Bifunctional Fluorescent Reagent]
CAS Number: 142246-48-8
MDL Number: MFCD00191405
Molecular Formula: C6H2Cl2N2O3S
Molecular Weight: 253.05
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 87


Other Product Names / Synonyms: 4-Chloro-2,1,3-benzoxadiazole-7-sulfonyl Chloride, 4-Chloro-7-chlorosulfonylbenzofurazan

Formula: C₆H₂Cl₂N₂O₃S
MW: 253.05 g/mol
Melting Pt: 87 °C
MDL Number: MFCD00191405
CAS Number: 142246-48-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e7582c94534cb7f8c5a39eb62a8de702c2af49c&vid=2e7582c94534cb7f8c5a39eb62a8de702c2af49c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR