(Chloromethyl)isopropoxydimethylsilane ≥96.0%

Supplier: TCI America

Synonyms: Chloromethyl-dimethyl-propan-2-yloxysilane

C1370-5ML C1370-25ML
TCC1370-005MLEA 73 USD
TCC1370-005ML TCC1370-25ML
(Chloromethyl)isopropoxydimethylsilane ≥96.0%
(Chloromethyl)isopropoxydimethylsilane

CAS Number: 18171-11-4
MDL Number: MFCD00010605
Molecular Formula: C6H15ClOSi
Molecular Weight: 166.72
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 28
Specific Gravity (20/20): 0.93


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (Chloromethyl)dimethylisopropoxysilane

Formula: C₆H₁₅ClOSi
MW: 166.72 g/mol
Boiling Pt: 145 °C
Density: 0.93 (20 °C)
Flash Pt: 28 °C
MDL Number: MFCD00010605
CAS Number: 18171-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad9822778c6f1d0883c0bc7017534609bd76790b&vid=ad9822778c6f1d0883c0bc7017534609bd76790b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR