Allyl methyl carbonate ≥98.0%

Supplier: TCI America

Synonyms: Carbonic acid Allyl Methyl Ester, Allyl Methyl Carbonate

C1315-25ML C1315-5ML
TCC1315-025MLEA 173 USD
TCC1315-025ML TCC1315-005ML
Allyl methyl carbonate ≥98.0%
Allyl methyl carbonate

CAS Number: 35466-83-2
MDL Number: MFCD00134545
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 131
Flash Point (°C): 33
Specific Gravity (20/20): 1.03


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Carbonic Acid Allyl Methyl Ester

Formula: C₅H₈O₃
MW: 116.1152 g/mol
Boiling Pt: 131 °C
Density: 1.03 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00134545
CAS Number: 35466-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6970f90464f55cb41b05744a0ba88cc183712ec5&vid=6970f90464f55cb41b05744a0ba88cc183712ec5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR