2-Aminopyridine-N-oxide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A0778-100MG A0778-1G A0778-5G
TCA0778-100MGEA 32 USD
TCA0778-100MG TCA0778-001G TCA0778-5G
2-Aminopyridine-N-oxide ≥98.0% (by GC, titration analysis)

CAS Number: 14150-95-9
MDL Number: MFCD00059168
Molecular Formula: C5H6N2O
Molecular Weight: 110.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 166

Formula: C₅H₆N₂O
Melting Pt: 166 °C
MDL Number: MFCD00059168
CAS Number: 14150-95-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50a1343c680879a9a1861f86620859bbd767d603&vid=50a1343c680879a9a1861f86620859bbd767d603&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR