Cumylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: alpha,alpha-Dimethylbenzylamine, Phenylisopropylamine, alpha-Aminoisopropylbenzene, 2-Phenylpropan-2-amine

C1293-5ML C1293-25ML
TCC1293-005MLEA 171.1 USD
TCC1293-005ML TCC1293-25ML
Cumylamine ≥98.0% (by GC, titration analysis)
Cumylamine

CAS Number: 585-32-0
MDL Number: MFCD00134680
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 197
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: alpha-Aminoisopropylbenzene, alpha,alpha-Dimethylbenzylamine, Phenylisopropylamine

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 197 °C
Density: 0.94 (20 °C)
MDL Number: MFCD00134680
CAS Number: 585-32-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c86bcb717c03383bc9e9e124e650b428cca9fd28&vid=c86bcb717c03383bc9e9e124e650b428cca9fd28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR