Allyl ethyl carbonate ≥97.0%
Supplier: TCI America
CAS Number: 1469-70-1
MDL Number: MFCD00049113
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 149
Specific Gravity (20/20): 0.99
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Carbonic Acid Allyl Ethyl Ester
Formula:
C₆H₁₀O₃ Boiling Pt: 149 °C Density: 0.99 (20 °C) |
MDL Number:
MFCD00049113 CAS Number: 1469-70-1 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...