Octanohydroxamic acid ≥99.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: N-Hydroxyoctanamide, Caprylohydroxamic acid

C1232-25G C1232-5G
TCC1232-025GEA 266 USD
TCC1232-025G TCC1232-5G
Octanohydroxamic acid ≥99.0% (by total nitrogen basis)
Octanohydroxamic acid

CAS Number: 7377-03-9
MDL Number: MFCD00143918
Molecular Formula: C8H17NO2
Molecular Weight: 159.23
Purity/Analysis Method: >99.0% (N)
Form: Crystal
Melting point (°C): 78


Other Product Names / Synonyms: Caprylohydroxamic Acid, N-Hydroxyoctanamide

Formula: C₈H₁₇NO₂
MW: 159.2261 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00143918
CAS Number: 7377-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0b6a7a789e98a99adf6d354ee187b4d023d0a92&vid=f0b6a7a789e98a99adf6d354ee187b4d023d0a92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR