(S)-(+)-1-Chloro-2-methylbutane ≥98.0%

Supplier: TCI America

C1188-10ML C1188-25ML
TCC1188-010MLEA 101 USD
TCC1188-010ML TCC1188-025ML
(S)-(+)-1-Chloro-2-methylbutane ≥98.0%
(S)-(+)-1-Chloro-2-methylbutane

CAS Number: 40560-29-0 MDL Number: MFCD00221494 Molecular Formula: C5H11Cl Molecular Weight: 106.59 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 100 Specific Gravity (20/20): 0.89 Specific rotation [a]20/D: 1.7 deg (neat) Storage Temperature: 0-10°C

Formula: C₅H₁₁Cl
MW: 106.59 g/mol
Boiling Pt: 100 °C
Density: 0.89 (20 °C)
MDL Number: MFCD00221494
CAS Number: 40560-29-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR