Isopropyl crotonate ≥98.0%

Supplier: TCI America

Synonyms: Crotonic Acid Isopropyl Ester, Isopropyl Crotonate

C1183-500ML C1183-25ML
TCC1183-500MLEA 684 USD
TCC1183-500ML TCC1183-025ML
Isopropyl crotonate ≥98.0%
Isopropyl crotonate

CAS Number: 6284-46-4
MDL Number: MFCD00059053
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Crotonic Acid Isopropyl Ester

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 147 °C
Density: 0.89 (20 °C)
MDL Number: MFCD00059053
CAS Number: 6284-46-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50949101fed44f88d08085878e7c9125c366fbe8&vid=50949101fed44f88d08085878e7c9125c366fbe8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR