(3-Chloro-2-hydroxypropyl)trimethylammonium chloride ∼65% in water
Supplier: TCI America
Synonyms:
3-Chloro-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride(65% in water), 3-Chloro-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride
C1180-25ML
C1180-500ML
TCC1180-025MLEA
21
USD
TCC1180-025ML
TCC1180-500ML
(3-Chloro-2-hydroxypropyl)trimethylammonium chloride ∼65% in water
(3-Chloro-2-hydroxypropyl)trimethylammonium chloride
CAS Number: 3327-22-8
MDL Number: MFCD00055655
Molecular Formula: C6H15Cl2NO
Molecular Weight: 188.09
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 110
Specific Gravity (20/20): 1.17
Formula:
C₆H₁₅Cl₂NO MW: 188.09 g/mol Density: 1.17 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00055655 CAS Number: 3327-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...