3-Chloropropyldimethoxymethylsilane ≥95.0%

Supplier: TCI America

Synonyms: 3-(Methyldimethoxysilyl)propyl Chloride, (3-Chloropropyl)dimethoxy(methyl)silane

C1168-5G C1168-25G
TCC1168-005GEA 18 USD
TCC1168-005G TCC1168-25G
3-Chloropropyldimethoxymethylsilane ≥95.0%
3-Chloropropyldimethoxymethylsilane

CAS Number: 18171-19-2
MDL Number: MFCD00053912
Molecular Formula: C6H15ClO2Si
Molecular Weight: 182.72
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Flash Point (°C): 49
Specific Gravity (20/20): 1.03


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: 3-(Methyldimethoxysilyl)propyl Chloride

Formula: C₆H₁₅ClO₂Si
MW: 182.72 g/mol
Boiling Pt: 185 °C
Density: 1.03 (20 °C)
Flash Pt: 49 °C
MDL Number: MFCD00053912
CAS Number: 18171-19-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4c02e6d4a96066cb58d624c918a7dd5021891c5&vid=b4c02e6d4a96066cb58d624c918a7dd5021891c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR