4-(4-ɑ-Cumylphenoxy)phthalonitrile ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
4-(4-(2-Phenylpropan-2-yl)phenoxy)phthalonitrile, 4-(4-α-Cumylphenoxy)phthalonitrile, 2-[4-(3,4-Dicyanophenoxy)phenyl]-2-phenylpropane
C1163-1G
C1163-5G
TCC1163-001GEA
141.2
USD
TCC1163-001G
TCC1163-005G
4-(4-ɑ-Cumylphenoxy)phthalonitrile ≥98.0% (by total nitrogen basis)
4-(4-alpha-Cumylphenoxy)phthalonitrile
CAS Number: 83482-57-9
MDL Number: MFCD00059048
Molecular Formula: C23H18N2O
Molecular Weight: 338.41
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 91
Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 2-[4-(3,4-Dicyanophenoxy)phenyl]-2-phenylpropane
Formula:
C₂₃H₁₈N₂O MW: 338.41 g/mol Melting Pt: 91 °C |
MDL Number:
MFCD00059048 CAS Number: 83482-57-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...