Direct Blue 86
Supplier: TCI America
Synonyms:
(Dihydrogenphthalocyaninedisulphonato)-coppe disodium salt
C1118-25G
C1118-100G
TCC1118-025GEA
42
USD
TCC1118-025G
TCC1118-100G
Direct Blue 86
Direct blue 86
CAS Number: 1330-38-7
MDL Number: MFCD00070592
Molecular Formula: C32H16CuN8O6S2
Molecular Weight: 780.16
Form: Crystal
Color: Deep Blue
Formula:
C₃₂H₁₆CuN₈O₆S₂ MW: 780.16 g/mol |
MDL Number:
MFCD00070592 CAS Number: 1330-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...