Tridecylamine ≥98.0% for synthesis

Supplier: TCI America
Danger

Synonyms: n-Tridecylamine, Tridecan-1-amine, 1-Aminotridecane

A0762-25G A0762-10G
TCA0762-25GEA 234.3 USD
TCA0762-25G TCA0762-010G
Tridecylamine ≥98.0% for synthesis
Tridecylamine

CAS Number: 2869-34-3
MDL Number: MFCD00008156
Molecular Formula: C13H29N
Molecular Weight: 199.38
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 275
Flash Point (°C): 110
Freezing point (°C): 27


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: n-Tridecylamine

Formula: H₃C(CH₂)₁₁CH₂NH₂
MW: 199.38 g/mol
Boiling Pt: 265
Melting Pt: 28.1
Density: 0.8049 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008156
CAS Number: 2869-34-3
EINECS: 220-694-0
UN: 3259
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64130a9f9b97dc4fcd9004fbfffdb2470a7f23a6&vid=d5dab8655a95a81e8c91c06167b29df637573166&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR