Carbamazepine ≥97.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
5H-Dibenzo[b,f]azepine-5-carboxamide
C1095-10G
C1095-25G
TCC1095-010GEA
60.24
USD
TCC1095-010G
TCC1095-25G
Carbamazepine ≥97.0% (by total nitrogen basis)
Carbamazepine
CAS Number: 298-46-4
MDL Number: MFCD00005073
Molecular Formula: C15H12N2O
Molecular Weight: 236.27
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 191
Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 5H-Dibenzo[b,f]azepine-5-carboxamide
Formula:
C₁₅H₁₂N₂O MW: 236.27 g/mol Melting Pt: 191 °C |
MDL Number:
MFCD00005073 CAS Number: 298-46-4 |
Specification Test Results
Purity/Analysis Method | >97.0% (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...