2-Chloroethyl chloroformate ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloroethyl carbonochloridate

C1052-25G C1052-100G
TCC1052-025GEA 43 USD
TCC1052-025G TCC1052-100G
2-Chloroethyl chloroformate ≥98.0%
2-Chloroethyl chloroformate

CAS Number: 627-11-2
MDL Number: MFCD00000646
Molecular Formula: C3H4Cl2O2
Molecular Weight: 142.96
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 154
Flash Point (°C): 60
Specific Gravity (20/20): 1.38
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Chloroformic Acid 2-Chloroethyl Ester

Formula: C₃H₄Cl₂O₂
MW: 142.9687 g/mol
Boiling Pt: 154 °C
Density: 1.38 (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00000646
CAS Number: 627-11-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d11753bc1e2cf00738c390cf943f7bcc3bdd58f6&vid=d11753bc1e2cf00738c390cf943f7bcc3bdd58f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR