Methyl-3-(4-methoxycarbonylphenyl)propionate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C1029-25G C1029-5G
TCC1029-25GEA 495.9 USD
TCC1029-25G TCC1029-005G
Methyl-3-(4-methoxycarbonylphenyl)propionate ≥98.0% (by titrimetric analysis)
Methyl-3-(4-methoxycarbonylphenyl)propionate

CAS Number: 40912-11-6
MDL Number: MFCD00059305
Molecular Formula: C12H14O4
Molecular Weight: 222.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 33
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-(4-Carboxyphenyl)propionic Acid Dimethyl Ester, 3-(4-Methoxycarbonylphenyl)propionic Acid Methyl Ester

Formula: C₁₂H₁₄O₄
Melting Pt: 33 °C
MDL Number: MFCD00059305
CAS Number: 40912-11-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a63bc238971daaf1a50d4335552496f3276c6da&vid=8a63bc238971daaf1a50d4335552496f3276c6da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR