Methyl-3-cyclohexene-1-carboxylate ≥98.0%
Supplier: TCI America
CAS Number: 6493-77-2
MDL Number: MFCD00053266
Molecular Formula: C8H12O2
Molecular Weight: 140.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Flash Point (°C): 60
Specific Gravity (20/20): 1.02
Other Product Names / Synonyms: 3-Cyclohexene-1-carboxylic Acid Methyl Ester
Formula:
C₈H₁₂O₂ Boiling Pt: 185 °C Density: 1.02 (20 °C) Flash Pt: 60 °C |
MDL Number:
MFCD00053266 CAS Number: 6493-77-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...