Cyclopentylmalonic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
C0945-5G
C0945-25G
TCC0945-5GEA
123
USD
TCC0945-5G
TCC0945-025G
Cyclopentylmalonic acid ≥98.0% (by titrimetric analysis)
Cyclopentylmalonic acid
CAS Number: 5660-81-1
MDL Number: MFCD00059515
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 162
Formula:
C₈H₁₂O₄ MW: 172.18 g/mol Melting Pt: 162 °C |
MDL Number:
MFCD00059515 CAS Number: 5660-81-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...