2-Chloroethyl ethyl sulfide ≥98.0%

Supplier: TCI America

C0938-25ML C0938-5ML
TCC0938-025MLEA 218 USD
TCC0938-025ML TCC0938-005ML
2-Chloroethyl ethyl sulfide ≥98.0%
2-Chloroethyl ethyl sulphide

CAS Number: 693-07-2
MDL Number: MFCD00000979
Molecular Formula: C4H9ClS
Molecular Weight: 124.63
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Flash Point (°C): 52
Specific Gravity (20/20): 1.07
Storage Temperature: 0-10°C

Formula: C₄H₉ClS
Boiling Pt: 157 °C
Density: 1.07 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00000979
CAS Number: 693-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f14518c8056a5b7e5d31cebc436eec2a504a9bc1&vid=f14518c8056a5b7e5d31cebc436eec2a504a9bc1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR