4-Allylmorpholine 97,0

Supplier: TCI America

Synonyms: N-allylmorpholine

A0756-5ML A0756-25ML
TCA0756-5MLEA 62 USD
TCA0756-5ML TCA0756-025ML
4-Allylmorpholine 97,0
4-Allylmorpholine

CAS Number: 696-57-1
MDL Number: MFCD00038034
Molecular Formula: C7H13NO
Molecular Weight: 127.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Specific Gravity (20/20): 0.93

Formula: C₇H₁₃NO
MW: 127.19 g/mol
Boiling Pt: 158 °C
Density: 0.93 (20 °C)
MDL Number: MFCD00038034
CAS Number: 696-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2c00b29c6fded1109abc928592fea23cd00a57&vid=ec2c00b29c6fded1109abc928592fea23cd00a57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR