α-Chlorothioanisole ≥95.0%

Supplier: TCI America

Synonyms: Chloromethyl phenyl sulphide

C0912-25G C0912-5G
TCC0912-25GEA 159 USD
TCC0912-25G TCC0912-5G
α-Chlorothioanisole ≥95.0%
α-Chlorothioanisole

CAS Number: 7205-91-6
MDL Number: MFCD00000921
Molecular Formula: C7H7ClS
Molecular Weight: 158.64
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 101
Specific Gravity (20/20): 1.23
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Chloromethyl Phenyl Sulfide

Formula: C₇H₇ClS
Density: 1.23 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00000921
CAS Number: 7205-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7ff734a3917194c515b4b2ce8dd0e3f621c3f4&vid=8bb12ff723525fd3d6a3d7459beb16c42467c5eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR