2-Chloro-3-ethylbenzoxazolium tetrafluoroborate ≥97.0% (by total nitrogen and titration analysis)
Supplier: TCI America
C0905-25G
C0905-5G
TCC0905-25GEA
225
USD
TCC0905-25G
TCC0905-005G
2-Chloro-3-ethylbenzoxazolium tetrafluoroborate ≥97.0% (by total nitrogen and titration analysis)
2-Chloro-3-ethylbenzoxazolium tetrafluoroborate
CAS Number: 63212-53-3
MDL Number: MFCD00011943
Molecular Formula: C9H9BClF4NO
Molecular Weight: 269.43
Purity/Analysis Method:
Formula:
C₉H₉BClF₄NO Melting Pt: 131 °C |
MDL Number:
MFCD00011943 CAS Number: 63212-53-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...