Atropine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (+/-)-Hyoscyamine, Tropine DL-tropate

A0754-5G A0754-25G
TCA0754-005GEA 150.5 USD
TCA0754-005G TCA0754-025G
Atropine ≥99.0% (by titrimetric analysis)
Atropine

CAS Number: 51-55-8
MDL Number: MFCD00022622
Molecular Formula: C17H23NO3
Molecular Weight: 289.38
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 116


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Neurotransmitter Agent Ingredients, Alkaloids (Phytochemicals), Tropane Alkaloids
Other Product Names / Synonyms: (+/-)-Hyoscyamine

Formula: C₁₇H₂₃NO₃
MW: 289.38 g/mol
Melting Pt: 116 °C
MDL Number: MFCD00022622
CAS Number: 51-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf77ac456b3bceae33e6cda79ea90dfcd9919b2d&vid=cf77ac456b3bceae33e6cda79ea90dfcd9919b2d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR