γ-Cyclodextrine ≥99.0% (by HPLC)
Supplier: TCI America
Synonyms:
gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
C0869-5G
C0869-25G
C0869-100G
TCC0869-005GEA
95.92
USD
TCC0869-005G
TCC0869-025G
TCC0869-100G
γ-Cyclodextrine ≥99.0% (by HPLC)
γ-Cyclodextrine
CAS Number: 17465-86-0
MDL Number: MFCD00009595
Molecular Formula: C48H80O40
Molecular Weight: 1297.13
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 178 deg (C=1, H2O)
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry, Bioscience, Oligosaccharides, Cyclodextrins
Other Product Names / Synonyms: Schardinger gamma-Dextrin, Cyclooctaamylose
Formula:
C₄₈H₈₀O₄₀ MW: 1297.1248 g/mol |
MDL Number:
MFCD00009595 CAS Number: 17465-86-0 EINECS: 241-482-4 |
Specification Test Results
Purity/Analysis Method | >99.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...