1,4-Cyclohexanediamine (cis and trans mixture) ≥97.0%

Supplier: TCI America

Synonyms: Cyclohexane-1,4-diamine, 1,4-Diaminocyclohexane

C0814-5ML C0814-25ML
TCC0814-005MLEA 26 USD
TCC0814-005ML TCC0814-025ML
1,4-Cyclohexanediamine (cis and trans mixture) ≥97.0%
1,4-Cyclohexanediamine (cis and trans mixture)

CAS Number: 3114-70-3
MDL Number: MFCD00035862
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 197
Flash Point (°C): 71


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 1,4-Diaminocyclohexane

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 197 °C
Flash Pt: 71 °C
MDL Number: MFCD00035862
CAS Number: 3114-70-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d509d58bda6573c5ece2bc7ab488ab03a04f04a&vid=2d509d58bda6573c5ece2bc7ab488ab03a04f04a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR