Hydroxypropyl acrylate (mixture of 2-hydroxypropyl and 2-hydroxy-1-methylethyl acrylate) ≥90.0% stabilized

Supplier: TCI America

Synonyms: Acrylic Acid Hydroxypropyl Ester, Propylene Glycol Monoacrylate

A0744-500ML A0744-25ML
TCA0744-500MLEA 48.83 USD
TCA0744-500ML TCA0744-025ML
Hydroxypropyl acrylate (mixture of 2-hydroxypropyl and 2-hydroxy-1-methylethyl acrylate) ≥90.0% stabilized
Hydroxypropyl acrylate (mixture of 2-hydroxypropyl and 2-hydroxy-1-methylethyl acrylate)

(stabilized with MEHQ)
CAS Number: 25584-83-2
MDL Number: MFCD04113589
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 77
Flash Point (°C): 100
Specific Gravity (20/20): 1.06


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid Hydroxypropyl Ester, Propylene Glycol Monoacrylate

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 77 °C
Density: 1.06 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD04113589
CAS Number: 25584-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5570ef14d4e929bbeff8387913412e272bff6d8d&vid=5570ef14d4e929bbeff8387913412e272bff6d8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR