2-Cyclohexylphenol ≥97.0%

Supplier: TCI America

C0732-25G C0732-5G
TCC0732-25GEA 92 USD
TCC0732-25G TCC0732-5G
2-Cyclohexylphenol ≥97.0%
2-Cyclohexylphenol

CAS Number: 119-42-6
MDL Number: MFCD00019335
Molecular Formula: C12H16O
Molecular Weight: 176.26
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 284
Melting point (°C): 56
Flash Point (°C): 134


Other Product Names / Synonyms: 2-Hydroxyphenylcyclohexane

Formula: C₁₂H₁₆O
Boiling Pt: 284 °C
Melting Pt: 56 °C
Flash Pt: 134 °C
MDL Number: MFCD00019335
CAS Number: 119-42-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e024e4b3ab0b0920928648c93eecc3e59864a01&vid=9e024e4b3ab0b0920928648c93eecc3e59864a01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR