N(α)-Cbz-DL-tryptophan ≥97.0%
Supplier: TCI America
Synonyms:
Cbz-DL-Trp-OH, Z-DL-Trp-OH
C0641-5G
C0641-1G
TCC0641-005GEA
75
USD
TCC0641-005G
TCC0641-001G
N(α)-Cbz-DL-tryptophan ≥97.0%
N(α)-Cbz-DL-tryptophan
CAS Number: 13058-16-7
MDL Number: MFCD00069707
Molecular Formula: C19H18N2O4
Molecular Weight: 338.36
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 170
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-DL-tryptophan, Z-DL-Trp-OH
Formula:
C₁₉H₁₈N₂O₄ Melting Pt: 170 °C |
MDL Number:
MFCD00069707 CAS Number: 13058-16-7 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...