Cyclohexylacetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Cyclohexaneacetic acid

C0589-25G C0589-250G
TCC0589-025GEA 88.9 USD
TCC0589-025G TCC0589-250G
Cyclohexylacetic acid ≥98.0% (by GC, titration analysis)
Cyclohexylacetic acid

CAS Number: 5292-21-7
MDL Number: MFCD00001518
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 113
Freezing point (°C): 30


Other Product Names / Synonyms: Cyclohexaneacetic Acid

Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 26 - 27 °C
Density: 0.9985 g/cm³ (22 °C)
Flash Pt: 113 °C
MDL Number: MFCD00001518
CAS Number: 5292-21-7
EINECS: 226-132-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5ae1e24b9d048648709ab37dfdb3c4ef3f4631b&vid=d5ae1e24b9d048648709ab37dfdb3c4ef3f4631b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR