N-Cyclohexyl-2-benzothiazolylsulfenamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

C0576-25G C0576-500G
TCC0576-025GEA 31.52 USD
TCC0576-025G TCC0576-500G
N-Cyclohexyl-2-benzothiazolylsulfenamide ≥98.0% (by HPLC, total nitrogen)
N-Cyclohexyl-2-benzothiazolylsulfenamide

CAS Number: 95-33-0
MDL Number: MFCD00022872
Molecular Formula: C13H16N2S2
Molecular Weight: 264.41
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 101

Formula: C₁₃H₁₆N₂S₂
Melting Pt: 101 °C
MDL Number: MFCD00022872
CAS Number: 95-33-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=615763ded08ded6decf46fdb4c101a685c72655a&vid=615763ded08ded6decf46fdb4c101a685c72655a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR