4-Chloropyrocatechol ≥98.0%

Supplier: TCI America

Synonyms: 4-Chloro-1,2-dihydroxybenzene, 4-Chlorobenzene-1,2-diol, 4-Chlorocatechol

C0568-25G C0568-5G
TCC0568-025GEA 600.5 USD
TCC0568-025G TCC0568-5G
4-Chloropyrocatechol ≥98.0%
4-Chloropyrocatechol

CAS Number: 2138-22-9
MDL Number: MFCD00059614
Molecular Formula: C6H5ClO2
Molecular Weight: 144.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 88


Other Product Names / Synonyms: 4-Chloro-1,2-dihydroxybenzene

Formula: C₆H₅ClO₂
MW: 144.56 g/mol
Boiling Pt: 140 °C
Melting Pt: 88 °C
MDL Number: MFCD00059614
CAS Number: 2138-22-9
EINECS: 218-381-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0467e5eb45ca0defae4fb208f5003d8ee21906b&vid=e0467e5eb45ca0defae4fb208f5003d8ee21906b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR