Sodium octanohydroxamate hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Octanohydroxamic Acid Sodium Salt
C0557-25G
C0557-1G
TCC0557-025GEA
706
USD
TCC0557-025G
TCC0557-001G
Sodium octanohydroxamate hydrate ≥98.0% (by titrimetric analysis)
Sodium octanohydroxamate hydrate
CAS Number: 50292-00-7
MDL Number: MFCD00058968
Molecular Formula: C8H17NO2
Molecular Weight: 181.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: Octanohydroxamic Acid Sodium Salt
Formula:
C₈H₁₇NO₂ MW: 181.21 g/mol |
MDL Number:
MFCD00058968 CAS Number: 50292-00-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...