DL-Cystine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (H-DL-Cys-OH)2

C0518-1G C0518-5G
TCC0518-001GEA 31 USD
TCC0518-001G TCC0518-5G
DL-Cystine ≥98.0% (by HPLC, titration analysis)
DL-Cystine

CAS Number: 923-32-0
MDL Number: MFCD00084652
Molecular Formula: C6H12N2O4S2
Molecular Weight: 240.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 260

Formula: C₆H₁₂N₂O₄S₂
Boiling Pt: 429 °C (1013 hPa)
Melting Pt: 260 °C
MDL Number: MFCD00084652
CAS Number: 923-32-0
EINECS: 213-094-5
Merck Index: 13,02811
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac9a0a228f0323b39bbba4bdf4f707c394055600&vid=ac9a0a228f0323b39bbba4bdf4f707c394055600&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR