L-Cysteic acid ≥98.0%

Supplier: TCI America

Synonyms: (R)-2-Amino-3-sulphopropanoic acid

C0514-25G C0514-1G
TCC0514-025GEA 155.44 USD
TCC0514-025G TCC0514-001G
L-Cysteic acid ≥98.0%
L-Cysteic acid

CAS Number: 498-40-8
MDL Number: MFCD00007524
Molecular Formula: C3H7NO5S
Molecular Weight: 169.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 8 deg (C=5, H2O)

Formula: C₃H₇NO₅S
MDL Number: MFCD00007524
CAS Number: 498-40-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67ee962b136012ae48dee70fc47b92ba184bbef9&vid=67ee962b136012ae48dee70fc47b92ba184bbef9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR