Cyclohexylhydrazine hydrochloride ≥98.0%

Supplier: TCI America

C0500-5G C0500-25G
TCC0500-005GEA 131 USD
TCC0500-005G TCC0500-25G
Cyclohexylhydrazine hydrochloride ≥98.0%
Cyclohexylhydrazine hydrochloride

CAS Number: 24214-73-1
MDL Number: MFCD00060160
Molecular Formula: C6H14N2
Molecular Weight: 150.65
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 112

Formula: C₆H₁₄N₂
Melting Pt: 112 °C
MDL Number: MFCD00060160
CAS Number: 24214-73-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=833b9f6287fdfaeffa382e8caafe5b196e859cc3&vid=833b9f6287fdfaeffa382e8caafe5b196e859cc3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR